Learn More
Deltalab™ Eurotubo Polypropylene Conical Tubes with Cap
Eurotubo Polypropylene Conical Tubes with Cap are sample tubes suitable for centrifugation tests in immunology, microbiology, etc. Centrifuge adapters are recommended to avoid tube malformations.
97.94€ - 165.36€
Specifications
Color | Clear |
---|---|
Material | Polypropylene |
For Use With | Centrifugation Tests in Immunology and Microbiology |
Packaging | Case |
Autoclavable | Autoclavable |
Description
- Samples and/or urine collection containers
- Available sterile or non-sterile
- Blue cap
Specifications
Clear | |
Centrifugation Tests in Immunology and Microbiology | |
Autoclavable |
Polypropylene | |
Case | |
Conical Tube |