Learn More
Deltalab™ Eurotubo Polypropylene Conical Tubes with Cap
Eurotubo Polypropylene Conical Tubes with Cap are sample tubes suitable for centrifugation tests in immunology, microbiology, etc. Centrifuge adapters are recommended to avoid tube malformations.
Brand: Deltalab™ 429910
This item is not returnable.
View return policy
Description
- Samples and/or urine collection containers
- Available sterile or non-sterile
- Blue cap
Specifications
Clear | |
Polypropylene | |
Case | |
Autoclavable | |
17 mm | |
5000 x g |
15 mL | |
Centrifugation Tests in Immunology and Microbiology | |
Non-sterile | |
122 mm | |
Conical Tube |