Learn More
DWK Life Sciences Wheaton™ Extended Controlled 20mm LDPE Dropper Tips
20mm extended controlled dropper tip made of LDPE (Low Density Polyethylene)
Brand: DWK Life Sciences W242438-A
This item is not returnable.
View return policy
Description
- Extended controlled dropper tip dispenses one drop at a time
- Average drop size is 44μL ±6μL
- Case of 100
Specifications
Yellow | |
1000 Cs. | |
41 μL | |
Plastic dropping bottle |
Polyethylene | |
ISO 9001 | |
Dropper Tip |