Learn More
SGE™ SilFlow™ GC 3 Port Splitter Kits
Highly efficient and reliable micro fluidic platform to improve GC connectivity.
Brand:  SGE™ 123722
750.20 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 13589000
  • 779.00€ / Each
Only null left
Estimated Shipment: 08-09-2025
Description
  • Kit includes a SilFlow microchannel device (MCD), SilTite FingerTite tool, mounting bracket, plus appropriate ferrules, nuts and blanking ferrules for set up
  • 1.1mm OD tubing not included
Specifications
Specifications
Port Splitter
0.25/0.32 mm
SilFlow microchannel device (MCD), SilTite FingerTite tool, Mounting bracket, Appropriately sized ferrules, nuts and blanking ferrules for set up
SGE Silflow Microchannel Device