Learn More
Nickel Electro™ Plastic Flat Lid
Reduce the evaporation and noise in an Ultrasonic Bath. Nickel Electro™ Clifton™ Plastic Flat Lid only requires one piece construction.
42.30€ - 203.00€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
13161323
|
Nickel Electro™
LD-1 |
Ultrasonic Bath SW1H |
42.30€
Each |
Estimated Shipment: 09-10-2025
Log in to see stock. |
|||||
13101333
|
Nickel Electro™
LD-30 |
Ultrasonic Bath SW30H |
203.00€
Each |
Estimated Shipment: 14-10-2025
Log in to see stock. |
|||||
13171323
|
Nickel Electro™
LD-3 |
Ultrasonic Bath SW3H |
47.90€
Each |
Estimated Shipment: 29-09-2025
Log in to see stock. |
|||||
13181323
|
Nickel Electro™
LD-6 |
Ultrasonic Bath SW6H |
56.35€
Each |
Estimated Shipment: 14-10-2025
Log in to see stock. |
|||||
13191323
|
Nickel Electro™
LD-12 |
Ultrasonic Bath SW12H |
183.00€
Each |
Estimated Shipment: 16-10-2025
Log in to see stock. |
|||||
Description
- One piece construction
- Reduces evaporation
- Reduces noise
- For use with Ultrasonic Bath
Specifications
Plastic |