Learn More
Nickel Electro™ Plastic Flat Lid
Reduce the evaporation and noise in an Ultrasonic Bath. Nickel Electro™ Clifton™ Plastic Flat Lid only requires one piece construction.
Brand: Nickel Electro™ LD-3
42.75 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- One piece construction
- Reduces evaporation
- Reduces noise
- For use with Ultrasonic Bath
Specifications
Ultrasonic Bath SW3H | |
Plastic |