Learn More
Copan Italia™ Non-Calibrated Loop
78.00€
Specifications
Product Type | Inoculating Loop |
---|---|
Gauge | 26 G |
Quantity | 25/Pk. |
Calibrated | No |
Product Code | Brand | Diameter (Metric) Inside | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Price | Quantity & Availability | |||||
11756528
|
Copan Italia™
311C25T |
3 mm |
78.00€
Pack of 25 |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
11766528
|
Copan Italia™
312C25T |
4 mm |
78.00€
Pack of 25 |
Estimated Shipment: 17-11-2025
Log in to see stock. |
|||||
11776528
|
Copan Italia™
313C25T |
5 mm |
78.00€
Pack of 25 |
Estimated Shipment: 17-11-2025
Log in to see stock. |
|||||
Specifications
Inoculating Loop | |
26 G | |
25/Pk. | |
No |