Learn More
Copan Italia™ Non-Calibrated Loop
Brand:  Copan Italia™ 313C25T
80.49 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11776528
  • 104.00€ / Pack of 25
Only null left
Estimated Shipment: 17-11-2025
Specifications
5 mm
26 G
No
Inoculating Loop
25/Pk.