Learn More
GRIN1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27676-1-AP-150UL
 View more versions of this product
Product Code. 16895765
  • 410.00€ / 150µL
Description
The GRIN1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GRIN1. This antibody recognizes human, mouse, rat antigen. The GRIN1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
GRIN1
Polyclonal
Unconjugated
150 μL
2902
IgG
Western Blot, Immunohistochemistry, ELISA
200 μg/mL
Rabbit
Primary
Human, Rat, Mouse