Learn More
behr Labor-Technik™ behrotest™ Dilution Water Tank
Made from low-density polyethylene. behr Labor-Technik™ behrotest™ Dilution Water Tank is designed to collect waste that is discharged directly from labs via special drain lines.
451.00€ - 1705.00€
Specifications
Color Blue
Material LDPE
Type Dilution Water Tank
View More Specs
Products 3
Product Code Brand Volume (Metric) Diameter (Metric) Includes Price Quantity & Availability  
Product Code Brand Volume (Metric) Diameter (Metric) Includes Price Quantity & Availability  
10460071
View Documents
behr Labor-Technik™
B00217796
30 L 350 mm Distributor frits, compressed air connection, internal stainless steel cooling hose and connection for recirculation thermostat
1705.00€
Each
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 24-09-2025
Add to basket
10516402
View Documents
behr Labor-Technik™
B00217792
30 L 350 mm Distributor frits, compressed air connection
451.00€
Each
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 22-09-2025
Add to basket
10065170
View Documents
behr Labor-Technik™
B00217793
60 L 400 mm Distributor frits, compressed air connection
566.00€
Each
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 22-09-2025
Add to basket
Description
Description
  • Includes aeration setup, level display and stop tap
  • Colored (light protection), with lid for easy cleaning
Specifications
Specifications
Blue
LDPE
Dilution Water Tank