Learn More
behr Labor-Technik™ behrotest™ Dilution Water Tank
Made from low-density polyethylene. behr Labor-Technik™ behrotest™ Dilution Water Tank is designed to collect waste that is discharged directly from labs via special drain lines.
Brand: behr Labor-Technik™ B00217792
311.62 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Includes aeration setup, level display and stop tap
- Colored (light protection), with lid for easy cleaning
Specifications
Blue | |
30 L | |
LDPE | |
Dilution Water Tank | |
350 mm |
Material: Low density polyethylene(LDPE) | |
Distributor frits, compressed air connection | |
Diameter: 350 mm, Height: 500 mm | |
500 mm |