Learn More
7-Fluoroisatin, 97%, Thermo Scientific Chemicals
CAS: 317-20-4 | C8H4FNO2 | 165.123 g/mol
28.44€ - 302.51€
Chemical Identifiers
CAS | 317-20-4 |
---|---|
Molecular Formula | C8H4FNO2 |
Molecular Weight (g/mol) | 165.123 |
MDL Number | MFCD01569508 |
InChI Key | HGBGVEOXPHGSOS-UHFFFAOYSA-N |
Synonym | 7-fluoroisatin, 7-fluoroindoline-2,3-dione, 7-fluoro-2,3-indolinedione, 1h-indole-2,3-dione, 7-fluoro, 7-fluoroindole-1h-2,3-dione, 7-fluoroindole-2,3-dione, 7-fluoro-2,3-dihydro-1h-indole-2,3-dione, 7-fluoroisatine, 7-fluoro-1h-benzo d azoline-2,3-dione, 7-fluor-1h-indol-2,3-dion |
PubChem CID | 586418 |
IUPAC Name | 7-fluoro-1H-indole-2,3-dione |
SMILES | C1=CC2=C(C(=C1)F)NC(=O)C2=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11326969
|
Thermo Scientific Alfa Aesar
H26827.03 |
1 g |
28.44€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11336969
|
Thermo Scientific Alfa Aesar
H26827.06 |
5 g |
90.75€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346969
|
Thermo Scientific Alfa Aesar
H26827.14 |
25 g |
302.51€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
7-Fluoroisatin is used as the intermediate of cardiovascular, anti-inflammatory and bactericidal drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications7-Fluoroisatin is used as the intermediate of cardiovascular, anti-inflammatory and bactericidal drugs.
Solubility
Sparingly Soluble in water (0.022 g/L)
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
317-20-4 | |
165.123 | |
HGBGVEOXPHGSOS-UHFFFAOYSA-N | |
586418 | |
C1=CC2=C(C(=C1)F)NC(=O)C2=O |
C8H4FNO2 | |
MFCD01569508 | |
7-fluoroisatin, 7-fluoroindoline-2,3-dione, 7-fluoro-2,3-indolinedione, 1h-indole-2,3-dione, 7-fluoro, 7-fluoroindole-1h-2,3-dione, 7-fluoroindole-2,3-dione, 7-fluoro-2,3-dihydro-1h-indole-2,3-dione, 7-fluoroisatine, 7-fluoro-1h-benzo d azoline-2,3-dione, 7-fluor-1h-indol-2,3-dion | |
7-fluoro-1H-indole-2,3-dione |
Specifications
317-20-4 | |
C8H4FNO2 | |
7-fluoroisatin, 7-fluoroindoline-2,3-dione, 7-fluoro-2,3-indolinedione, 1h-indole-2,3-dione, 7-fluoro, 7-fluoroindole-1h-2,3-dione, 7-fluoroindole-2,3-dione, 7-fluoro-2,3-dihydro-1h-indole-2,3-dione, 7-fluoroisatine, 7-fluoro-1h-benzo d azoline-2,3-dione, 7-fluor-1h-indol-2,3-dion | |
HGBGVEOXPHGSOS-UHFFFAOYSA-N | |
7-fluoro-1H-indole-2,3-dione | |
586418 | |
97% |
193°C to 195°C | |
MFCD01569508 | |
Sparingly Soluble in water (0.022g/L) | |
C1=CC2=C(C(=C1)F)NC(=O)C2=O | |
165.123 | |
165.12 | |
7-Fluoroisatin |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures