Learn More
7-Fluoroisatin, 97%, Thermo Scientific Chemicals
Product Code. 11336969
Click to view available options
Quantity:
1 g
5 g
25 g
CAS: 317-20-4 | C8H4FNO2 | 165.123 g/mol
7-Fluoroisatin is used as the intermediate of cardiovascular, anti-inflammatory and bactericidal drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
7-Fluoroisatin is used as the intermediate of cardiovascular, anti-inflammatory and bactericidal drugs.
Solubility
Sparingly Soluble in water (0.022 g/L)
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 317-20-4
Molecular Formula C8H4FNO2
Molecular Weight (g/mol) 165.123
MDL Number MFCD01569508
InChI Key HGBGVEOXPHGSOS-UHFFFAOYSA-N
Synonym 7-fluoroisatin, 7-fluoroindoline-2,3-dione, 7-fluoro-2,3-indolinedione, 1h-indole-2,3-dione, 7-fluoro, 7-fluoroindole-1h-2,3-dione, 7-fluoroindole-2,3-dione, 7-fluoro-2,3-dihydro-1h-indole-2,3-dione, 7-fluoroisatine, 7-fluoro-1h-benzo d azoline-2,3-dione, 7-fluor-1h-indol-2,3-dion
PubChem CID 586418
IUPAC Name 7-fluoro-1H-indole-2,3-dione
SMILES C1=CC2=C(C(=C1)F)NC(=O)C2=O
Specifications
Melting Point 193°C to 195°C
Quantity 5 g
Solubility Information Sparingly Soluble in water (0.022g/L)
Formula Weight 165.12
Percent Purity 97%
Chemical Name or Material 7-Fluoroisatin