Learn More
Tenak™ 40 mm Cryobox Dividers
Dividers for use with 75 mm high cryoboxes.
67.75€ - 77.95€
Specifications
Type | Grid Divider |
---|---|
For Use With (Equipment) | 75 mm high cryoboxes |
Height (Metric) | 40 mm |
Product Code | Brand | Holds | Tube Array | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Tube Array | Price | Quantity & Availability | ||||
15240454
|
Tenak™
TE22132 |
Vials with 15 mm O.D. | 8 x 8 cells |
73.15€
Pack of 36 |
Estimated Shipment: 02-09-2025
Log in to see stock. |
||||
15210634
|
Tenak™
TE22707 |
Vials with 15 mm O.D. | 6 x 6 cells |
70.80€
Pack of 36 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||
15220554
|
Tenak™
TE22137 |
Vials with 10 mm O.D. | 12 x 12 cells |
77.95€
Pack of 36 |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
15220634
|
Tenak™
TE22708 |
Vials with 25.25 mm O.D. | 5 x 5 cells |
70.80€
Pack of 36 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||
15230634
|
Tenak™
TE22709 |
Vials with 31.75 mm O.D. | 4 x 4 cells |
70.80€
Pack of 36 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||
15240634
|
Tenak™
TE22710 |
Vials with 42.50 mm O.D. | 3 x 3 cells |
70.80€
Pack of 36 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||
15280544
|
Tenak™
TE22124 |
Vials with 41 mm O.D. | 3 x 3 cells |
67.75€
Pack of 36 |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
15290544
|
Tenak™
TE22125 |
Vials with 31 mm O.D. | 4 x 4 cells |
67.75€
Pack of 36 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||
11521785
|
Tenak™
TE22705 |
Vials with 15.50 mm O.D. | 8 x 8 cells |
75.55€
Pack of 36 |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
11531785
|
Tenak™
TE22706 |
Vials with 17.75 mm O.D. | 7 x 7 cells |
75.55€
Pack of 36 |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
11571685
|
Tenak™
TE22130 |
Vials with 17 mm O.D. | 7 x 7 cells |
73.15€
Pack of 36 |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
11591685
|
Tenak™
TE22134 |
Vials with 13 mm O.D. | 9 x 9 cells |
73.15€
Pack of 36 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||
15200554
|
Tenak™
TE22126 |
Vials with 24 mm O.D. | 5 x 5 cells |
67.75€
Pack of 36 |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||
Description
- For use in cardboard cryoboxes
- Holds cryogenic vials
- Grid style divider
Specifications
Grid Divider | |
75 mm high cryoboxes | |
40 mm |