Learn More
Zirconyl Chloride Octahydrate, 98%, Reagent grade, Honeywell™
Product Code. 15602240 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Poly Bottle
Reagent Grade, 98%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13520-92-8
Molecular Formula Cl2H16O9Zr
Molecular Weight (g/mol) 322.24
MDL Number MFCD00149898
InChI Key SDFOCNSOSARAKC-UHFFFAOYSA-L
Synonym zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402
PubChem CID 159678
IUPAC Name oxozirconium;octahydrate;dihydrochloride
SMILES O.Cl[Zr](Cl)=O
Specifications
Color White
Quantity 100 g
Linear Formula ZrOCl2 · 8H2O
UN Number UN3260
Formula Weight 322.25g/mol
Percent Purity 98%
Grade Reagent
Packaging Poly Bottle
Appearance Powder or crystals
Density 1.91g/cm3
Chemical Name or Material Zirconyl Chloride Octahydrate