Learn More
SNV Comines™ Xavier™ Cotton Laboratory Coats
Provides general light protection against splashes in the laboratory. SNV Comines™ Xavier™ Cotton Laboratory Coats are available in a range of sizes.
25.00€
Specifications
Product Type | Xavier Lab Coat |
---|---|
Gender | Men's |
Material | Carbon |
Color | White |
Closure Type | Snaps |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
12651126
|
SNV Comines™
XAVLP00400T1 |
40 |
25.00€
Each |
In Stock
Log in to see stock. ![]() |
|||||
12671126
|
SNV Comines™
XAVLP00400T2 |
44 |
25.00€
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||||
Description
- White in color
- With snap buttons
- Collared with lapels
Specifications
Xavier Lab Coat | |
Carbon | |
Snaps | |
2 Flat pockets, 1 breast pocket, 1 inside pocket, opening angle |
Men's | |
White | |
4 | |
100% Cotton |