Learn More
Haleco™ NeutraBase Wood Sodium Phosphate/Cellulose
For neutralizing and solidifying bases for easy and safe cleaning.
Brand: Haleco™ JLN100B
450.43 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Makes the spill gel together and decreases the pH
- Color indicator changes from white to violet during the neutralization, then returns to white once the neutralization has finished
- Non-toxic
- Incinerable
- Ideal for the chemical industry and laboratories
- Provided in a plastic bucket
Specifications
White to violet | |
9.55 kg | |
9.55 kg |