Learn More
DWK Life Sciences Wheaton™ Septa for Open-Top Caps
Shop All Wheaton Science Products Products
Click to view available options
Septum:
PTFE-faced natural rubber
PTFE-faced silicone
Closure Size:
15 mm
20 mm
22 mm
33 mm
Unit Size:
Pack of 100
Description
- For caps with open tops
- Autoclavable
Specifications
Specifications
Material | PTFE, Rubber |
Sterility | Sterile |
Septum | PTFE-faced natural rubber |
Autoclavable | Autoclavable |
Type | Septa |
Closure Size | 20 mm |