Learn More
DWK Life Sciences Wheaton™ Micro Kit and Accessories
WHEATON standard taper Clear-Seal™ grease-free joints provide smooth, unground surfaces, which seal without grease and reduce seizing and contamination from grease
82.00€ - 109.00€
Specifications
| For Use With (Equipment) | Wheaton micro kit |
|---|---|
| Includes | 19/22 Clear-Seal™ joints |
| Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
|
12485432
|
DWK Life Sciences
123753 |
Connecting Adapter |
82.00€
Each |
Estimated Shipment: 28-11-2025
Log in to see stock. |
|||||
|
12455432
|
DWK Life Sciences
109933 |
Claisen Adapter |
109.00€
Each |
Estimated Shipment: 28-11-2025
Log in to see stock. |
|||||
Description
- Apparatus for a wide range of organic chemistry experiments at a lower price than purchasing the components separately
- Clear-Seal™ joints can replace ground units in any application and provide other important advantages over ground joints
- Superior in vacuum and radioactive applications
- Smooth surfaces resist contamination
- Cleaning is simple and thorough
- Interchangeable with ground joints
- All chemical changes and thermometer scales remain visible through the joints
- Clear-Seal joints have never been weakened by grinding and are formed from special wall tubing that is heavier than standard wall for extra strength
- Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to ASTM E 438 Type I, Class A requirements
- Clear-Seal joints are produced to American Standard Taper dimensions and can be used interchangeably with any standard taper ground joints
Specifications
| Wheaton micro kit | |
| 19/22 Clear-Seal™ joints |