Learn More
DWK Life Sciences Wheaton™ Black Phenolic Screw Caps with PTFE Faced 14B White Rubber Liner
Provides completely inert inner seal and surface for sample or product
235.00€ - 468.00€
Specifications
Product Type | Screw Cap |
---|---|
Liner Material | TFE, Styrene Butadiene Rubber |
Autoclavable | Autoclavable |
Material | Phenolic |
Color | Black |
Product Code | Brand | Closure Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Price | Quantity & Availability | |||||
11511235
|
DWK Life Sciences
W240406 |
8-425 |
235.00€
Pack of 200 |
In Stock
Log in to see stock. |
|||||
12416171
|
DWK Life Sciences
W242711 |
24-400 |
468.00€
Pack of 200 |
Estimated Shipment: 12-12-2025
Log in to see stock. |
|||||
Description
- Autoclavable
Specifications
Screw Cap | |
Autoclavable | |
Black |
TFE, Styrene Butadiene Rubber | |
Phenolic |