Learn More
DWK Life Sciences Wheaton™ Amber Boston Rounds with Polyvinyl-lined Caps
Amber Type III soda lime glass protects light-sensitive solutions
Brand: DWK Life Sciences W216837
39.28 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Amber Type III soda lime glass protects light-sensitive solutions
- Available in partitioned Convenience Pack or Valu-Bulk™ cartons
- Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage
Each case of Convenience Pack bottles includes Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage
Order Info
Caps can be ordered separately.
Compliance
USP Type III soda-lime glass
Specifications
2 oz., 60 mL | |
2 oz. | |
1.53 in. | |
Convenience pack | |
94 mm | |
Poly/Vinyl | |
20-400 mm | |
Screw Cap |
60 mL | |
Amber | |
Soda Lime Glass | |
White | |
3.7 in. | |
Polypropylene | |
39 mm |