Learn More
DWK Life Sciences Wheaton™ 2D Data Matrix Bar Code Bottom Insert
For use with Wheaton CryoELITE™ Freestanding Cryogenic Vials
Brand:  DWK Life Sciences W224621-0303
88.88 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12379959
  • 91.15€ / Pack of 100
Only null left
Estimated Shipment: 23-12-2025
Description
  • Makes sample identification and traceability reliable and convenient
  • Bar code insert manually press fits into bottom of vial
  • Data matrix readable with standard scanning hardware
  • Made from same cryogenic radiation-grade, virgin polypropylene as vial, ensuring uniform thermal expansion
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene
Sterile
500/Cs.