Learn More
Cytiva Whatman™ Potassium Iodide Test Paper on a Reel
Combines ease-of-use with unsurpassed accuracy and consistency
Brand: Cytiva 2602-500A
21.18 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This rapid determination of pH values has led to many applications in laboratories and industry
- Instant pH readings
- Accurate for a wide range of routine pH testing
- Inexpensive
- Convenient and portable for field use
Specifications
For detecting chlorine and other oxidizing agents | |
Each | |
Potassium Iodide Test Paper |
Safety and Handling
- reel, lead acetate paper dispenser
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1A
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : acetic acid, lead(2+) salt, trihydrate