Learn More
Socorex™ Acura™ 8-channel Micropipette Volumetric Module
Increase the versatility of your Acura™ electronic micropipette control unit and save costs by accessorizing it with one of 27 available volumetric modules. Socorex™ 8-channel Micropipette Volumetric Module is an excellent and easily attachable/detachable choice for your multi-channel micropipetting needs.
559.00€ - 600.00€
Specifications
For Use With | 8 Channel Pipette |
---|---|
Number of Channels | 8 |
Product Line | Acura |
Type | Micropipette Volumetric Module |
For Use With (Application) | 8 Channel Pipette |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10234762
|
Socorex™
800.08.350 |
350 μL |
559.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10682373
|
Socorex™
800.08.050 |
50 μL |
559.00€
Each |
Estimated Shipment: 27-10-2025
Log in to see stock. |
|||||
10315732
|
Socorex™
800.08.010 |
10 μL |
600.00€
Each |
Estimated Shipment: 27-10-2025
Log in to see stock. |
|||||
Description
- One of 27 available volumetric assemblies.
- Increase versatility and save costs.
- Disassembles and reassembles without tools.
- Calibration specificities of extra modules - as set by factory QC or by pipette owner - are retained in instrument memory.
Specifications
8 Channel Pipette | |
Acura | |
8 Channel Pipette | |
1 Each |
8 | |
Micropipette Volumetric Module | |
Acura™ Electro 926 Micropipette |