Learn More
Fisherbrand™ Stainless Steel Offset Spoons
The unique curved shape spoon, offset, allows contact with all corners of bottles and containers reducing sample losses. The alternate end can be used as a scraper to chip away at hard samples or used as a spatula for sampling or lifting.
Brand:  Fisherbrand™ 8205
Product Code. 11533492
  • 12.25€ / Each
Only null left
Estimated Shipment: 16-07-2025
Add to basket
Description
Description
  • Laboratory Offset Spoon
  • Stainless steel 304 spec 18/8 non-magnetic, bright finish
  • Uses the finest in corrosion-resistant 304 stainless steel
  • The unique curved shape spoon, offset, allows contact with all corners of bottles and containers reducing sample losses 
  • The alternate end can be used as a scraper to chip away at hard samples or used as a spatula for sampling or lifting
  • Can be used within laboratory to retrieve powders, granules, solids and viscous liquids
Specifications
Specifications
1 Ea.
Autoclavable
Bright
-196°C to +550°C
190 mm
Sterile
Laboratory Offset Spoon
For Sterile Applications
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions