Learn More
3M™ Versaflo™ Battery TR-830/94243(AAD)
3M™ Versaflo™ Battery TR-830/94243(AAD) is a lithium-ion battery pack designed for use with the 3M™ Versaflo™ TR-800 Series Powered Air Turbo.
Brand: 3M™ 7100150925
681.48 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Charges quickly (90 % charge in approximately 3.5 hours)
- Approximately 750 full charge/discharge cycles
- Standard capacity rechargeable lithium ion battery
- Mechanical attachment of battery to help prevent battery removal and spark risk during use per EN 60079-11
- Built in charge level indicator
- Intrinsically safe
Specifications
6 to 10 hr | |
CE Approved, NIOSH Approved | |
TR-830 |
Lithium-ion | |
Battery |