Learn More
VACUUBRAND™ Vacuubrand Operating Module C3-B For VACUU·BUS™ Plug Connection
The upgrade kit SYNCHRO for a second vacuum connection can be equipped, depending on the application, with a manual flow control valve (677137) or an in-line solenoid valve (636668) for electronic vacuum control with the CVC 3000 vacuum controller. These funtional elements for individual vacuum control are necessary to operate the vacuum ports.
Brand:  VACUUBRAND™ 20636668
549.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10576855
  • 555.00€ / Each
Only null left
Estimated Shipment: 12-08-2025
Specifications
For VACUU·BUS™ Plug Connection
VACUU·BUS™ Plug
1 Each
Operating Module C3-B