Learn More
UVP Mini Ultraviolet Lamp Replacement Tube
Replacement tubes for lightweight, compact and financial UV Lamps
30.00€ - 48.20€
Specifications
Wattage 4 W
Packaging Quantity 1 Ea.
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
11796832
View Documents
UVP
34-0010-01
Model UVL-4, UVL-23R handheld UV lamp and mini UV lamp
30.00€
Each
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 26-09-2025
Add to basket
11788231
View Documents
UVP
34-0004-01
Model UVGL-15 and UVSL-14P compact and mini handheld UV lamp
48.20€
Each
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 26-09-2025
Specifications
4 W