Learn More
DuPont™ Tyvek 400 Dual
Provides an ideal balance of protection, durability and comfort. DuPont™ Tyvek™ 400 Dual Hooded Coverall is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres down to 1 micron in size.
Brand: DuPont™ D14809637
805.31 EUR valid until 2025-09-30
Use promo code "24380" to get your promotional price.
Description
- Coverall with 3-piece hood
- Tyvek™ at the front and large breathable SMS back
- Stitched external seams
- Elasticated wrists, ankles and face
- Elasticated waist (stitched-in)
- Tyvek™ zipper and flap
- White
- Antistatic
Specifications
Hooded Coveralls | |
Tyvek, SMS | |
Elastic | |
Elastic | |
Elastic | |
Stitched | |
(EU) 2016/425, EN 1149-5 |
X-Large | |
White | |
Elastic | |
Hooded | |
Zipper with Storm Flap | |
Category III, Type 5 and 6 | |
108 to 116 cm |