Learn More
Emma Safety Footwear Treviso Safety Shoes
The foot-hugging fit provides the wearer with a naturally light and airy on-foot feel. Water resistant uppers and a woven insole make the shoes virtually penetration proof. The 100% nitrile rubber outsole repels liquids, harmful or otherwise, and provides extra grip on almost all surfaces.
Brand: Emma Safety Footwear MM10509241
107.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- O3-Work shoe without steel toecap with sole plate
- Full grain leather construction
- Nitrile Rubber Heat Resistant Outsole
- Metal-free
- Chrome-free lining
- ESD certified
- Sanitized
Specifications
EN ISO 20347:2012 | |
Black | |
Hydro-Tec™ Sanitized Silver & Wellness leather | |
41 |
Laces | |
Treviso | |
Leather |