Learn More
Titanium Nickel powder composite, -325 mesh, Thermo Scientific Chemicals
35.10€ - 835.16€
Specifications
Chemical Name or Material | Titanium Nickel powder composite |
---|---|
Avg. Mol. Wt. or Mol. Wt. Range | Ti:Ni; 70:30 wt% |
MDL Number | MFCD02091734 |
UN Number | UN3089 |
Solubility Information | Insoluble in water. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11329259
|
Thermo Scientific Alfa Aesar
088384.09 |
10 g | N/A | ||||||
11349259
|
Thermo Scientific Alfa Aesar
088384.22 |
100 g |
233.16€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11339259
|
Thermo Scientific Alfa Aesar
088384.36 |
500 g |
835.16€
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A 70% titanium, 30% nickel composite powder strained through -325 mesh. Also used in alloy powder, bone staples and antioxidant coating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Titanium Nickel powder composite | |
MFCD02091734 | |
Insoluble in water. | |
Air Sensitive |
Ti:Ni; 70:30 wt% | |
UN3089 | |
−325 Mesh | |
Powder |
Safety and Handling
GHS H Statement
H228-H351-H317
Flammable solid.
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P210-P240-P241-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c
H228-H317-H351
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures