Learn More
Terumo™ Three-Part Syringe
Use these syringes for the aspiration of fluids or sampling blood or other body fluids. Terumo™ Three-Part Syringes feature a unique double beveled design for greater patient comfort.
Brand: Terumo™ 8SS02S21381
This item is not returnable.
View return policy
Description
- Ultra-sharp needles
- Clear barrels
- Bold, precise scale markings
- Smooth plunger action
- Tamper evident peel packs
Specifications
Autoclavable | |
21 G | |
40 mm | |
100/Pk. | |
Sterile |
Polypropylene | |
Yes | |
3-part Syringe | |
2 mL |