Learn More
Thermo Scientific™ Chromacol™ Storage Vials
Click to view available options
Color:
Clear
Unit Size:
Pack of 187
Description
- Use caps with standard GPI threads
- Class I hydrolytic extraction neutral borosilicate glass
- May be used with injection caps with piercable seals, or solid PTFE lined storage caps
Specifications
Specifications
Color | Clear |
Diameter (Metric) Outer | 15 mm |
Material | Borosilicate Glass |
Volume (Metric) | 5.0 mL |
Thread Style | Custom External |
Volume (English) | 1.35 dr. |
Closure Type | Screw Cap |
Height (Metric) | 48 mm |