Learn More
Texwipe™ TexBond™ 20 Cleanroom Bond Paper
Cleanroom bond paper with exceptional contamination control characteristics needed in critical environments. It has been especially formulated to address the needs of users requiring a paper that retains low ion and particle levels with significantly improved copying and writing qualities.
20.85€ - 21.00€
Specifications
Dimensions (L x W) | 210 x 297 mm |
---|---|
Quantity | 250 Sheets/Pk., 10 Packs/Cs. |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
11525415
|
Texwipe™
TX5711 |
Blue |
21.00€
Pack of 250 |
Estimated Shipment: 22-12-2025
Log in to see stock. |
|||||
16405029
|
Texwipe™
TX5719 |
White |
20.85€
Pack of 250 |
Estimated Shipment: 29-09-2025
Log in to see stock. |
|||||
Description
- Possesses both strength and excellent heat resistance, making it ideal for use in laser printers and photocopiers
- Very low particle generation
- Very low ion levels
- High opacity rating
- Precision cut edges
- Cleanroom packaged Benefits
- Exceptional copying ability
- Laser printer and photocopier compatible
- Clear reproductions
- Autoclavable for use in sterile environments
- Photocopying, ink jet and laser printing
- Offset printing
- Cleanroom manuals, work instructions, note taking and data transfer
- Color coded carriers
Specifications
210 x 297 mm | |
250 Sheets/Pk., 10 Packs/Cs. |