Learn More
Sampling Systems™ TeleScoop Water Sampler
The TeleScoop water sampler is an extendable sampler ideal for waters and liquids in hard-to-reach places. Its extendable rods reach up to 6 m in length, and the robust yet lightweight design means easy handling, even for the longest of rods.
25.15€ - 93.30€
Specifications
Product Type | Telescopic Rod |
---|---|
Material | Aluminum |
Autoclavable | Not Autoclavable |
Graduated | No |
Sterility | Non-sterile |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
17962112
|
Sampling Systems™
5065A-1001 |
1000 mm |
31.35€
Each |
Estimated Shipment: 06-08-2025
Log in to see stock. |
|||||
17922032
|
Sampling Systems™
5065A-1201 |
1200 mm |
25.15€
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||||
17942032
|
Sampling Systems™
5065A-2501 |
2500 mm |
31.35€
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||||
17809366
|
Sampling Systems™
5065A-2801 |
2800 mm |
51.75€
Each |
Estimated Shipment: 25-08-2025
Log in to see stock. |
|||||
17952032
|
Sampling Systems™
5065A-3001 |
3000 mm |
51.75€
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
17962032
|
Sampling Systems™
5065A-4501 |
4500 mm |
67.35€
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||||
17972032
|
Sampling Systems™
5065A-6001 |
6000 mm |
93.30€
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||||
Description
- Dual locking system and multi angle adaptor elbow so that numerous attachments can be preset to angles for effective sampling
- Attachments are quickly and easily changed without the need for using additional tools
- Variety of compatible sampling accessories
- Ideal for sampling from river banks, overhead gantries, etc.
Specifications
Telescopic Rod | |
Not Autoclavable | |
Non-sterile |
Aluminum | |
No |