Learn More
Sulfuric Acid, 0.005M, Chem Lab™
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
Specifications
Specifications
Appearance | Colorless |
pH | <2 |
Composition | Sulfuric acid 0.005 mol/L |
Concentration | 0.491g H2SO4/l H2O = 0.01 N (±0.00002/20°C) |
Density | 1g/mL |
Odor | Odorless |
Quantity | 1 L |
Physical Form | Liquid |
Solubility Information | Soluble in Water |