Learn More
Cole-Parmer™ Stuart™ Heater Block Insert
Alumium heating blocks for block heaters
Brand: Cole-Parmer™ 04805-76
This item is not returnable.
View return policy
Description
- Safest way to heat round-bottom flasks
- Flexible modular system for a wide range of sizes up to 500 mL
- Low-well design is perfect for multi-neck flasks
Specifications
Anodized Aluminum | |
Block | |
base SHB250BASE | |
Medium |