Learn More
Cole-Parmer™ Stuart™ Gabled Lid
Enhance the performance of your Techne baths and circulators
Brand:  Cole-Parmer™ 12122-95
405.52 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12747969
  • 434.00€ / Each
Only null left
Estimated Shipment: 17-09-2025
Description
  • Lids, trays, temperature probes, and other accessories for your bath.
  • Manufactured from stainless steel and available to fit 8 L baths to help prevent evaporation losses.
Specifications
Specifications
8L Bath