Learn More
Cole-Parmer™ Stuart™ Flat Lid
Enhance the performance of your Techne baths and circulators
155.00€ - 244.00€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
12717969
|
Cole-Parmer™
12122-89 |
12L Bath |
231.00€
Each |
Estimated Shipment: 13-11-2025
Log in to see stock. |
|||||
12727969
|
Cole-Parmer™
12122-90 |
18 and 26L Bath |
187.00€
Each |
In Stock
Log in to see stock. |
|||||
12737969
|
Cole-Parmer™
12122-94 |
48L Bath |
244.00€
Each |
Estimated Shipment: 13-11-2025
Log in to see stock. |
|||||
12707969
|
Cole-Parmer™
12122-88 |
8L Bath |
155.00€
Each |
Estimated Shipment: 13-11-2025
Log in to see stock. |
|||||
Description
Lids, trays, temperature probes, and other accessories for your bath.
Manufactured from stainless steel and available to fit 8 L baths to help prevent evaporation losses.