Learn More
Sampling Systems™ SteriWare™ Disposable Powder Funnel
Constructed of chemically resistant, food grade HDPE. Sampling Systems™ SteriWare™ Disposable Powder Funnel is ideal for transferring and sampling powders.
Brand: Sampling Systems™ 8111H-250
81.21 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- No cross contamination
- No cleaning required
Specifications
250 mm | |
HDPE | |
No | |
Filling Funnel (Powders) |
40 mm | |
Not Autoclavable | |
Non-sterile |