Learn More
Bel-Art™ Sterileware™ Flat Bottom Sampling Scoop
Flat bottom sampling scoops are used by pharmaceutical, food, cosmetic and chemical companies for general production or laboratory work.
259.00€ - 376.74€
Specifications
Product Type | Scoop |
---|---|
Material | High-impact Polystyrene |
Autoclavable | Not Autoclavable |
Disposable | Single-use |
Sterility | Sterile |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
16612814
|
Bel-Art™
F36911-0025 |
25 mL |
260.73€
Pack of 100 |
Estimated Shipment: 21-10-2025
Log in to see stock. |
|||||
16622814
|
Bel-Art™
F36911-0050 |
50 mL |
293.99€
Pack of 100 |
Estimated Shipment: 21-10-2025
Log in to see stock. |
|||||
16632814
|
Bel-Art™
F36911-0100 |
100 mL | |||||||
16642814
|
Bel-Art™
F36911-0150 |
150 mL | |||||||
16652814
|
Bel-Art™
F36911-0250 |
250 mL |
357.92€
Pack of 100 |
Estimated Shipment: 21-10-2025
Log in to see stock. |
|||||
16662814
|
Bel-Art™
F36911-0500 |
500 mL |
351.08€
Pack of 40 |
Estimated Shipment: 21-10-2025
Log in to see stock. |
|||||
16672814
|
Bel-Art™
F36911-1000 |
1000 mL |
376.74€
Pack of 30 |
Estimated Shipment: 21-10-2025
Log in to see stock. |
|||||
Description
- Flat bottom allows for use as a weigh boat
- Approximate volumes can be measured by removing excess powder
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High impact polystyrene (HIPS) scoops heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10-6)
- Ready for one time use and disposal
- Lot stamped for accurate tracing
- Shipped with a Certificate of Processing for Sterilization
Specifications
Scoop | |
Not Autoclavable | |
Sterile |
High-impact Polystyrene | |
Single-use |