Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SN55, 15 mm
Non-perforated slide-in drip trays constructed of stainless steel.
Brand:  Memmert™ SN55B02072
84.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17578675
  • 87.15€ / Each
Only null left
Estimated Shipment: 01-10-2025
Description
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
Specifications
SN55 Sterilizer
Drip Tray
Rim: 15 mm