Learn More
Bel-Art™ Stainless-steel Sampling Spoon and Spatula
Ends coated with brown Fluo-Kem™ PTFE FEP.
Brand: Bel-Art™ H36711-0000
Description
Ends coated with brown Fluo-Kem™ PTFE FEP.
- Coating provides anti-stick, corrosion-resistant surface
- Rod flattened at one end to serve as spatula with spoon on opposite end
Specifications
Spoon and Spatula Sampler | |
Not Autoclavable | |
Brown Fluo-Kem PTFE FEP |
Stainless Steel | |
4mm dia. rod flattened at one end to serve as 8 x 51mm spatula with 13 x 25mm spoon on opposite end | |
22.9 cm |