Learn More
RUBIS High Precision Tweezers
High Precision Tweezers - Antimagnetic/Anti-acid SS
18.73€ - 36.75€
Specifications
Material | Anti-acid, Anti-magnetic Stainless Steel |
---|---|
Product Type | High Precision Tweezers |
Quantity (No. per Case) | Each |
Product Code | Brand | Design | Tip Style | Length (Metric) | Length (English) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Design | Tip Style | Length (Metric) | Length (English) | Price | Quantity & Availability | ||
11713426
![]() |
RUBIS
3 |
Straight | Pointed, Ultra Fine | 120 mm | 4.72 in. |
20.63€
Each |
In Stock
Log in to see stock. |
||
11783256
![]() |
RUBIS
00 |
Straight | Pointed, Thick | 115 mm | 4.52 in. |
20.23€
Each |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||
11783266
![]() |
RUBIS
FTP |
Straight | Conical, Ultra Fine | 120 mm | 4.72 in. |
30.74€
Each |
Estimated Shipment: 12-08-2025
Log in to see stock. |
||
11743266
![]() |
RUBIS
SS |
Straight | Pointed, Ultra Fine | 135 mm | 5.31 in. |
19.33€
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||
11849133
![]() |
RUBIS
OC11 |
Straight | Pointed, Ultra Fine | 110 mm | 4.32 in. |
18.73€
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||
11773266
![]() |
RUBIS
0C9 |
Straight | Pointed, Ultra Fine | 90 mm | 3.54 in. |
19.83€
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||
11733266
![]() |
RUBIS
H |
Straight, Strong, Sturdy | Fine | 90 mm | 3.54 in. |
20.23€
Each |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||
11723426
![]() |
RUBIS
3L |
Straight, Self Locking | Pointed, Ultra Fine | 120 mm | 4.72 in. |
36.75€
Each |
Estimated Shipment: 30-07-2025
Log in to see stock. |
||
11733426
![]() |
RUBIS
2AL |
Straight, Self-Locking | Round, Flat | 120 mm | 4.72 in. |
28.85€
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
||
Description
- High Precision Tweezers
- 100% Made in Switzerland
- Anti-acid/Antimagnetic Stainless Steel
- Perfect alignment
- Very effective
- Reliable
- Excellent chemical resistance
Specifications
Anti-acid, Anti-magnetic Stainless Steel | |
High Precision Tweezers | |
Each |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title