Learn More
DWK Life Sciences Stainless Steel Handle, for staining tray
Shop All DWK Life Sciences ProductsDescription
Microscopy is a method of viewing objects that lie outside the resolution of the human eye and can be examined. The objects to be observed are magnified to such an extent that they can be displayed as images. For sample preparation, the objects to be examined are cut into very thin slices. These sections are then applied to an object slide and covered with a cover glass and appropriate solutions. To view the sections under the microscope, the sections on the slide must be filled in a staining trough and stained with an appropriate staining solution.
- Can be attached separately and not permanently to the dye rack, making it easy to insert and remove the slides
- Allows the staining rack to be safely placed in the staining box while direct contact with the staining solution can be avoided
Specifications
Specifications
Autoclavable | Not Autoclavable |
Material | Stainless Steel |
For Use With (Application) | Transport of staining trays; Suspension of the dye rack |
No. per Pack | 10/Pk. |
Quantity | 10/Pk. |