Learn More
Nickel Electro™ Clifton™ Stainless Steel Gable Lids
Obtain optimum results keeping samples in a water bath free from contamination and saving energy. Designed allowing for full use of whole width of a Clifton™ water bath, Clifton™ Stainless Steel Gable Lids help reduce energy consumption and running costs.
Brand:  Nickel Electro™ SL1-4
186.30 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11429519
  • 194.00€ / Each
Only null left
Estimated Shipment: 29-09-2025
Description
  • Designed allowing full use of whole width of a Clifton™ water bath
  • Helps reduce evaporation
  • Asists in keeping samples in a water bath free from contamination and saves energy
  • Allows extended use of the bath, right into the edges of the tank
  • Lid design ensures all condensate returns back into the bath
  • Insulated handles for safe use
  • Reduces energy consumption and running costs for your Clifton water bath
  • Lowest height of the gable being 45mm above the bath
Specifications
Specifications
2 and 4 L Unstirred Baths