Learn More
Nickel Electro™ Stainless Steel Funnels
This durable stainless steel funnel, break-resistant, stain proof and easy to clean makes it an ideal choice for most laboratory and research environments.
Brand:  Nickel Electro™ FUNSS080H
Product Code. 12953621
  • 12.10€ / Each
Only null left
Estimated Shipment: 22-07-2025
Description
  • Laboratory Funnel, with handle, stainless steel 304 spec 18/8 non-magnetic, bright finish
  • Durable stainless steel funnel, break-resistant, stain proof and easy to clean
  • May be used with or without filter membranes for gravity filtration as well as simple funnel procedures
  • Flute is vented to allow complete, uninhibited flow
  • Can be used with liquids or solids
  • The handle can also be used as a convenient hanging point when not in use
  • Smooth finish on internal surfaces making funnel easy to clean or sterilise (sterilize)
Specifications
Specifications
Stainless Steel
Vented flute