Learn More
OHAUS™ Stainless Steel Flask Clamp
OHAUS™ Stainless Steel Flask Clamp is designed to securely grip and position various sizes of flasks.
37.45€ - 48.25€
Specifications
Type | Flask Clamp |
---|---|
Material | Stainless Steel |
Color | Silver |
Jaw Type | Four Interlocking |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15762499
|
OHAUS™
30400084 |
10 mL Flask |
37.50€
Each |
Estimated Shipment: 22-10-2025
Log in to see stock. |
|||||
15772499
|
OHAUS™
30400085 |
25 mL Flask |
37.50€
Each |
In Stock
Log in to see stock. |
|||||
15782499
|
OHAUS™
30400086 |
50 mL Flask |
37.45€
Each |
In Stock
Log in to see stock. |
|||||
15792499
|
OHAUS™
30400087 |
125 mL Flask |
37.45€
Each |
In Stock
Log in to see stock. |
|||||
15702509
|
OHAUS™
30400088 |
250 mL Flask |
42.75€
Each |
In Stock
Log in to see stock. |
|||||
15712509
|
OHAUS™
30400089 |
500 mL Flask |
48.25€
Each |
In Stock
Log in to see stock. |
|||||
Description
- Constructed of stainless steel
- For use with flasks
- Designed to securely hold flask in place
Specifications
Flask Clamp | |
Stainless Steel | |
Silver | |
Four Interlocking |