Learn More
Grant Instruments™ Stainless Steel Bath Cover
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Bath Cover for Grant Stirred Water Baths is available flat or gabled with removable hinge.
108.00€ - 692.00€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
12757779
|
Grant Instruments™
STL12 |
ST12 tank |
382.00€
Each |
Estimated Shipment: 07-11-2025
Log in to see stock. |
|||||
12787779
|
Grant Instruments™
STL5 |
ST5 tank |
108.00€
Each |
In Stock
Log in to see stock. |
|||||
12777779
|
Grant Instruments™
STL38 |
ST38 tank |
692.00€
Each |
In Stock
Log in to see stock. |
|||||
Description
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Bath Cover for Grant Stirred Water Baths is available flat or gabled with removable hinge.
- Stainless steel construction
- Plastic handle
- Available flat (STL 5), or gabled with removable hinge (STL 12, STL 38)
- For use with Grant Stirred Water Baths and Heated Circulators
- Prevents heat from escaping
- Reduces sample contamination
Specifications
Stainless Steel |