Learn More
Kartell™ Staining Jar with 2 Lids
Click to view available options
Quantity:
4/Pk.
Unit Size:
Pack of 4
Description
- Moulded in PMP
- Supplied with 2 lids
- One lid for use when dish is not in use, providing evaporation free storage of staining liquid
- The other lid with special opening to allow handle of staining rack to be inserted
- Xylene can cause etching after 2 days at room temperture
Specifications
Specifications
Closure Included | Closure Included |
Closure Material | PMP |
Bottom Shape | Flat |
Color | Clear |
Material | PMP |
Type | Staining Jar |
Quantity | 4/Pk. |
Dimensions (L x W) | 95 x 74 mm |
Length (Metric) | 74 mm |
Width (Metric) | 95 mm |